Learn More
Thermo Scientific™ Barnstead™ MegaPure™ ACS
Product Code. 12743848 Shop All Thermo Scientific Products
Click to view available options
Type:
Automatic Collection System
Wall-mount Bracket
For Use With (Equipment):
Automatic Collection System
Mega-Pure Glass Still
Unit Size:
Each
Product Code. 12743848
Brand: Thermo Scientific™ 400634
Only null left
Add to basket
Add to basket
Control the operation of your Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Still with this automatic collection system, which collects up to 45L of water.
Control the operation of your glass still with the Thermo Scientific™ Barnstead™ MegaPure™ ACS (Automatic Collection System), which collects up to 45L of water.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Automatic Collection System
Type Wall-mount Bracket
Quantity Each