missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Access Port

Product Code. 30238332 Shop All BINDER Products
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30238332

Brand: BINDER™ 80122499

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.

  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600

Specifications

For Use With (Equipment) KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Material Silicone
Type Access Port
Ports 1 on Top
Port Size 30 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.