Learn More
BINDER™ Base with Casters
For Use With: CB 220
Brand:  BINDER™ 9051-0029
Additional Details : Weight : 61.00000kg
Product Code. 15682206
  • 590.00€ / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • For use with CB Series Carbon Dioxide Incubators
  • Includes casters
Specifications
Specifications
Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator
Accessory, Base with Casters
Casters