Learn More
Memmert™ Basic Series Water Baths
Product Code. p-8001878 Shop All Memmert Products
Click to view available options
Capacity (Metric):
7 L
10 L
14 L
22 L
29 L
45 L
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 10003832
Brand: Memmert™ WNB45
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Controller Type Microprocessor PID
Capacity (Metric) 45 L
Certifications/Compliance CE, VDE, GS
Temperature Range +10°C to +95°C
Display Digital LED
For Use With (Application) For research laboratories, food and beverage, temperature control
Material Stainless Steel
Model WNB 45
Temperature (Metric) Maximum 95°C
Temperature Resolution ±0.1°C
Length (Metric) 818 mm
Length (Metric) Interior 590 mm
Width (Metric) 516 mm
Width (Metric) Interior 350 mm
Height (Metric) 296 mm
Height (Metric) Interior 220 mm
Depth (Metric) Interior 590 mm
Shipping Weight (Metric) 31 kg
Weight (Metric) 24 kg
Voltage 230 V
Wattage 2.8 kW
Frequency 50/60 Hz