Learn More
BD BBL™ Crystal™ ID Systems Accessories
Product Code. 11768912
Click to view available options
Product Type:
Anaerobic Nh inoculum
Calibration Standard
Microbiology Media
Quantity:
10 Pack
Each
Unit Size:
Each
Pack of 10
Product Code. 11768912
Brand: BD 245038
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
BBL Crystal MIND Software
  • Interprets the test results and provides specimen identification
  • Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
Codebooks Include Enteric/Nonfermenter, Anaerobe, Neisseria/Haemophilus, Gram-Positive, Rapid Gram-Positive) Supplied in individual hard-copy binder BBL Crystal Panel Viewer
  • For facilitated viewing and recording of chromogenic and fluorogenic reactions
BBL CrystalSpec™ Standards†
  • Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard
†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type Anaerobic Nh inoculum
For Use With (Equipment) Crystal™ ID system
Quantity 10 Pack