Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
Brand: BD 221716
Additional Details : Weight : 0.01940kg
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP | |
15°C to 25°C | |
United States | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L | |
Size K tubes with cap |
Beige | |
Liquid | |
Trypticase Soy Broth | |
For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms | |
100 x 5 mL |
For in vitro diagnostic use.