Learn More
Brady™ BBP12 Label printer 300 dpi - EU with Cutter, Unwinder and Brady Workstation LAB Suite
Product Code. 17107011
Click to view available options
:
Resolution: 300 dpi
Regulatory: This product is under eco-contribution tax.
Product Code. 17107011
Brand: Brady™ 306867
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
A compact, reliable, affordable entry level thermal transfer benchtop printer.
  • The Brady BBP™12 label printer is a compact, reliable, affordable entry level thermal transfer benchtop printer.
  • Ideal for customers requiring low volume printing on lots of different materials within the electrical, voice and datacom, LAB and specialized electronics markets.
  • It allows you to print labels, tapes cable markers, sleeves and much more.
  • The printer comes with the Brady Workstation Laboratory Identification Software suite enabling you to immediately start designing professional labels.
  • This suite combines frequently used software apps to create laboratory identification labels.
Specifications
Display Type Color LCD
Print Technology Thermal Transfer
Width (Metric) 202.00 mm
Height (Metric) 173.00 mm
Depth (Metric) 258.00 mm
Dimensions (L x W x H) 258.00 x 202.00 x 173.00 mm
Color Capability Monochrome
Includes Pre-installed cutter, ships with an unwinder, USB Cable, power cord, product CD, Quick Start Guide and Brady Workstation Laboratory ID Suite
Print Speed 101.60 mm/s
Resolution 300 dpi
Power Supply Details AC Adaptor
Alternate Part Number BBP12-EU-U-C-LABS