missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 (Follicular Lymphoma Marker) Mouse, Clone: rBCL6/1527, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 510.00€
Specifications
Antigen | Bcl-6 (Follicular Lymphoma Marker) |
---|---|
Clone | rBCL6/1527 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920808
|
EnquireBio
604-MSM10-P1 |
100 μg |
510.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910808
|
EnquireBio
604-MSM10-P0 |
20 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
Bcl-6 (Follicular Lymphoma Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human bcl-6 protein fragment (apprx. aa256-389) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
95kDa |
rBCL6/1527 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
604 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Nuclear | |
Raji or Ramos cells. Tonsil or Hodgkin's lymphoma. |