Description
They are practical for note taking and data recording in the cleanroom. Bond is made with the purest quality cellulose from specially selected sustainable forests and is coated with a proprietary formula to minimise particle and fibre release.
Specifications
Specifications
| Material | Cellulose |
| Ruled | College Ruled |
| Certifications/Compliance | ISO Class 3 - 7, Cleanroom Class 1 - 10,000, EU Grade B/C/D |
| Packaging | 50 Sheets/book |
| Size | A4 |