missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: AK Agar No.2 (Sporulating Agar)

Product Code. 10373395
Click to view available options
:
Arret and Kirshbaum (AK) Agar No. 2; 500g
This item is not returnable. View return policy

Product Code. 10373395

Brand: BD 210912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For isolation, detection and cultivation microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Arret and Kirshbaum Agar No. 2
Product Type Agar
Formulation Pancreatic digest of Gelatin 6g/L, Pancreatic digest of Casein 4g/L, Yeast extract 3g/L, Beef extract 1.5g/L, Dextrose 1g/L, Agar 15g/L, Manganous Sulfate 0.3g/L
For Use With (Application) For preparation of spore suspensions for use in procedures for the detection of antibiotic residues in Milk and dairy products
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.