missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: PPLO Broth Base without Crystal Violet

Product Code. 11768123
Click to view available options
:
Mycoplasma Broth Base; 500g
This item is not returnable. View return policy

Product Code. 11768123

Brand: BD 211458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mycoplasma Broth Base for isolation of micro-organisms

TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated
Media Type Mycoplasma Broth Base
Product Type Microbiology Media
Formulation Beef heart infusion from (solids) 2g/L, Pancreatic digest of Casein 7g/L, Beef extract 3g/L, Yeast extract 3g/L, Sodium Chloride 5g/L
For Use With (Application) Antimicrobial susceptibility testing and isolation of microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.