missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Oxgall
Click to view available options
:
Oxgall; BD Difco; 500g
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | <30°C |
| Additive | Oxgall |
| Form | Powder |
| For Use With (Application) | Selection of Bile tolerant bacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P330-Rinse mouth.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
- MIXTURE LIST-Contains : Oxgall
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction