Learn More
BD BBL™/Difco™ TB Fluorescent Stain Kit M
Product Code. 15191866
Click to view available options
Quantity:
4 Pack
Unit Size:
Each
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15191866
Brand: BD 212519
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Includes
One 250mL bottle each: Auramine M, Decolorizer and Potassium Permanganate
For staining mycobacteria by the Morse, Blair, Weiser and Sproat fluorescent procedure
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
TRUSTED_SUSTAINABILITY
Specifications
Type TB Fluorescent Stain Kit
Quantity 4 Pack
Includes Auramine M, decolorizer, and potassium permanganate
Format Bottle
For Use With (Application) For staining mycobacteria by the Morse, Blair, Weiser and Sproat fluorescent procedure
Storage Requirements 15° to 30°C
Packaging Type Bottle
Stain Type Fluorescent Stain Kit M (Auramine M, Decolorizer, Potassium Permanganate)
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Kit Tb Fluorescent Stain Kit M
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P235-Keep cool.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use water spray, fog, CO2, dry chemical or alcohol resistant foam to extinguish.
  • P403-Store in a well-ventilated place.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol, phenol,4,4'-carbonimidoylbis[N,N-dimethylaniline]
For in vitro Diagnostic Use.