Learn More
Biosciences Transcription Factor Buffer Set, BD
Click to view available options
Quantity:
Ea.
Unit Size:
Each
Description
Transcription Factor Buffer Set was designed to improve ease-of-use and minimize processing time, to reduce nonspecific staining, to increase the resolution of positively stained cells and to significantly reduce cell loss during fixation, permeabilization and staining procedures.
- Contents: Fixation/permeabilization buffer, diluent buffer, perm/wash buffer
Specifications
Specifications
| Chemical Name or Material | Transcription factor Buffer set |
| For Use With (Application) | Intracellular block, flow cytometry, staining of transcription factors alone or in combination with cell surface markers and cytokines |
| Quantity | Ea. |
Safety and Handling
Product Identifier
- BD Pharmingen Transcription Factor Buffer Set
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : formaldehyd e...%, methanol