missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse MIP-1β; Flex Set, Bead C9

Product Code. 15859318
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15859318 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15859318 Brand BD Brand No. 558343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Mouse MIP-1β; Standard; Mouse MIP-1β; PE Detection Reagent; Mouse MIP-1β; Capture Bead C9

For detecting mouse Macrophage Inflammatory Protein-1 in serum and cell culture samples

The BD CBA Mouse MIP-1β Flex Set is a bead-based immunoassay capable of measuring mouse Macrophage Inflammatory Protein-1β (MIP-1β), also known as CCL4, in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse MIP-1β Flex Set.

  • Reactivity: Mouse
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1beta
Cross Reactivity Mouse
Includes Mouse MIP-1beta, standard, Mouse MIP-1beta, PE detection reagent, Mouse MIP-1beta, capture bead C9
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Mouse MIP-1beta flex set (bead C9)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.