missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: APT Broth
Click to view available options
:
Agar All-purpose (APT) Broth with Tween 80; 500g
Specifications
Specifications
| Certifications/Compliance | COMPF, USDA |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | APT Agar |
| Product Type | Microbiology Media |
| Formulation | Yeast extract 7.5g/L, Pancreatic digest of Casein 12.5g/L, Dextrose 10g/L, Sodium Citrate 5g/L, Thiamine Hydrochloride 1mg/L, Sodium Chloride 5g/L, Dipotassium Phosphate 5g/L, Manganese Chloride 0.14g/L, Magnesium Sulfate 0.8g/L, Ferrous Sulfate 0.04g/L, |
| For Use With (Application) | Cultivation of heterofermentative lactoBacilli and other organisms requiring high thiamine content |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction