Learn More
BD Difco™ Dehydrated Culture Media: B12 Inoculum Broth
Click to view available options
:
B12 Inoculum Broth USP; 100g
Description
- Formulation: Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Dipotassium Phosphate 2g/L, Polysorbate 80 1g/L
Certifications
USP
Specifications
Specifications
| Certifications/Compliance | USP |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | B12 Inoculum Broth |
| Product Type | Microbiology Media |
| Formulation | Tomato Juice 5g/L, Proteose Peptone No.3 7.5g/L, Yeast extract 7.5g/L, Dextrose 10g/L, Dipotassium Phosphate 2g/L, Polysorbate 80 1g/L |
| For Use With (Application) | Cultivation of wide variety of microorganisms |
| Packaging Type | Bottle |
| Quantity | 100 g |