missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: DRBC Agar (Dichloran Rose Bengal Chloramphenicol)
Click to view available options
:
DRBC Agar (Dichloran Rose Bengal Chloramphenicol); 500g
Specifications
Specifications
| Certifications/Compliance | BAM, CCAM, COMPF, SMD |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | DRBC Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone No. 3 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Dichloran 2mg/L, Rose Bengal 25mg/L, Chloramphenicol 0.1g/L, Agar 15g/L |
| For Use With (Application) | Supports good growth of Yeasts and molds while inhibiting bacteria and the spreading of rapidly growing molds |
| Packaging Type | Bottle |
| Quantity | 500 g |
Safety and Handling
Signal Word
- Danger
- Carcinogenicity Category 1A
- H350-May cause cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : chloramphenicol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction