missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Micro Inoculum Broth
Click to view available options
:
Micro Inoculum Broth; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Micro Inoculum Broth |
| Product Type | Microbiology Media |
| Formulation | Proteose Peptone 5g/L, Yeast extract 20g/L, Dextrose 10g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 0.1g/L |
| For Use With (Application) | Preparing the inoculum of lactoBacilli and other microorganisms used in microbiological assays of vitamins and amino acid |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction