missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Wilkins-Chalgren Agar
Click to view available options
:
Wilkins-Chalgren Agar; 500g
Description
- Formulation: Pancreatic digest of Casein 10g/L, Peptone 10g/L, Yeast extract 5g/L, Dextrose 1g/L, Sodium Chloride 5g/L, L-Arginine 1g/L, Sodium Pyruvate 1g/L, Hemin 5mg/L, Vitamin K1 0.5mg/L, Agar 15g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
ISO/DIS, 11014CCAM
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014CCAM |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Wilkins-Chalgren Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 10g/L, Peptone 10g/L, Yeast extract 5g/L, Dextrose 1g/L, Sodium Chloride 5g/L, L-Arginine 1g/L, Sodium Pyruvate 1g/L, Hemin 5mg/L, Vitamin K1 0.5mg/L, Agar 15g/L |
| For Use With (Application) | Susceptibility testing of anaerobes and for isolating and cultivating anaerobes |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction