missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: XLT4 Agar Base
Click to view available options
:
XLT4 Agar Base; 500g
Description
- Formulation: Proteose Peptone No. 3 1.6g/L, Yeast extract 3g/L, L-Lysine 5g/L, Xylose 3.75g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Sodium Chloride 5g/L, Agar 18g/L, Phenol red 0.08g/L
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
ISO, 9000, USDA
Specifications
Specifications
| Certifications/Compliance | ISO, 9000, USDA |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | XLT4 Agar Base |
| Product Type | Agar |
| Formulation | Proteose Peptone No. 3 1.6g/L, Yeast extract 3g/L, L-Lysine 5g/L, Xylose 3.75g/L, Lactose 7.5g/L, Saccharose 7.5g/L, Ferric Ammonium Citrate 0.8g/L, Sodium Thiosulfate 6.8g/L, Sodium Chloride 5g/L, Agar 18g/L, Phenol red 0.08g/L |
| For Use With (Application) | Isolation of Enterococcus faecalis 29212, Escherichia coli 25922, Proteus mirabilis 25933, Salmonella enterica subspecies enterica serotype Typhimurium 14028, Staphylococcus aureus 25923 |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction