missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Listeria O Antisera

Used as positive control in identification of Listeria monocytogenes in the slide agglutination test

Brand:  BD 223021

Additional Details : Weight : 0.00100kg


 View more versions of this product

Product Code. 11732764

  • 417.00€ / 1mL
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Listeria monocytogenes is a widespread problem in public health and the food industries.

  • Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
  • Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4

For in vitro Diagnostic Use.

Certifications

CE marked

Specifications

Specifications

CE Marked; For in vitro diagnostic use
Antiserum
1 mL
Refrigerate, store at 2°C to 8°C
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documents

Documents

Certificates
Promotions

Promotions