Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks
Product Code. p-4902080
Click to view available options
Type:
EZ Incubation Rack
Holds:
Petri Dishes (15 to 18)
Petri Dishes (30 to 33)
Unit Size:
Each
Product Code. 11707204
Brand: BD 260673
Only null left
Add to basket
Add to basket
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
  • Holds 15-18 Petri dishes
TRUSTED_SUSTAINABILITY
Specifications
Type EZ Incubation Rack
Format Box Rack
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Holds Petri Dishes (15 to 18)
Product Line GasPak
Quantity Each
Size Standard