missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human Lineage Cell Depletion Set - DM
Click to view available options
Quantity:
5 mL
Unit Size:
Each
Description
Includes
Biotin Mouse Anti-human CD3, Biotin Mouse Anti-human CD14, Biotin Mouse Anti-human CD16, Biotin Mouse Anti-human CD19,Biotin Mouse Anti-human CD41a,Biotin Mouse Anti-human CD56, Biotin Mouse Anti-human Glycophorin A, Streptavidin Particles Plus - DM
For depletion of mature hematopoietic cells from PBMC, bone marrow, cord blood samples
- Contains biotinylated monoclonal antibodies
- Enriches rare cell populations such as stem cell, progenitor cells, dendritic cells
- Contains sufficient reagents to label 1 x 10e9 peripheral blood mononuclear cells
- Streptavidin Particles Plus - DM are magnetic nanoparticles
Applications:
Cell separation
Specifications
Specifications
| Sample Type | Bone Marrow, Cord Blood, PBMC |
| Type | Human Lineage Cell Depletion Set |
| For Use With (Application) | Cell separation |
| Includes | Biotin Mouse Anti-Human CD3, Biotin Mouse Anti-Human CD14, Biotin Mouse Anti-Human CD16, Biotin Mouse Anti-Human CD19, Biotin Mouse Anti-Human CD41a, Biotin Mouse Anti-Human CD56, Biotin Mouse Anti-Human Glycophorin A, Streptavidin particles Plus - DM, Biotinylated monoclonal Antibodies |
| Quantity | 5 mL |
For research use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction