missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse IFN-γ (AN-18) ELISA Set

Product Code. 15826718
Click to view available options
Quantity:
20 Plates
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15826718

missing translation for 'mfr': BD 551866

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Capture Antibody: Anti-mouse IFN-Γ monoclonal antibody (clone AN-18)
  • Detection Antibody: Biotinylated anti-mouse IFN-Γ monoclonal antibody
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
  • Standards: Recombinant mouse IFN-Γ, lyophilized
  • Instruction / Analysis Certificate

For Immunoassay protocols

The OptEIA Set for mouse interferon-Γ (IFN-Γ), utilizing clone AN-18, contains components necessary to develop enzymelinked immunosorbent assays.

  • Reactivity: Mouse
  • For natural or recombinant mouse IFN-Γ in serum, plasma, and cell culture supernatants

Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse IFN-gamma (AN-18) ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Mouse IFN-g in Serum, Plasma and cell culture supernatants
Quantity 20 Plates
Storage Requirements 2°C to 8°C

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.