Learn More
Perm Buffer IV (10x), BD
Click to view available options
Quantity:
50 mL
Unit Size:
Each
Description
BD Phosflow™ Perm Buffer IV (Perm Buffer IV) is intended for the permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells (PBMC) and lysed whole blood cells.
- Treatment with buffer and subsequent washing with staining buffer enables access for fluorescent antibodies to stain intracellular molecules
- Buffer preserves light scattering properties of cells
- Providies capacity for intracellular immunofluorescent staining in preparation for Phosflow cytometric analysis
Flow Cytometry, Intracellular staining
Specifications
Specifications
| Chemical Name or Material | Cell Permeabilization Buffer |
| Concentration | 10X |
| For Use With (Application) | Permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells and lysed whole blood cells, flow cytometry, intracellular staining |
| Quantity | 50 mL |