Learn More
Plus Permeabilization Buffer, BD
Click to view available options
Quantity:
10 mL
Unit Size:
Each
Description
- Used as a staining enhancer and secondary permeabilization reagent
- Should be used with fixed cell samples only
Flow Cytometry, Intracellular staining
Specifications
Specifications
| Chemical Name or Material | Permeabilization Buffer |
| For Use With (Application) | Immunofluorescent staining of incorporated BrdU for flow cytometric analysis, staining enhancer and secondary permeabilization reagent, intracellular staining |
| Quantity | 10 mL |