Learn More
BD Ratiometric Calcium Assay Kit
Involves single dye addition step with no subsequent washing
Brand:  BD 644243
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15131975
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Includes: Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
Description
Description
  • For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
Specifications
Specifications
-20°C
1 vial ratiometric calcium indicator, 10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer
Detection Assay Kit
Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels
10 Preps