Learn More
BD Ratiometric Calcium Assay Kit
Involves single dye addition step with no subsequent washing
Brand:  BD 644243
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15131975
  • 665.00€ / Pack of 10
Estimated Shipment
03-05-2021
Add to basket
Includes: Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
Description
Description
  • For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
Specifications
Specifications
Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels
10 Preps
Ratiometric Calcium Assay Kit
1 vial ratiometric calcium indicator, 10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer
-20°C