missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Ratiometric Calcium Assay Kit
Involves single dye addition step with no subsequent washing
739.35€
Specifications
Content And Storage | -20°C |
---|---|
For Use With (Application) | Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels |
Type | Detection Assay Kit |
Includes: Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
15131975
|
BD
644243 |
1 vial ratiometric calcium indicator, 10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer | 10 Preps |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
- For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
Specifications
-20°C | |
Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels | |
Detection Assay Kit |