Learn More
BD Urine Preservative Transport
Description
Includes
Each Qx UPT Kit contains 100 Qx UPT tubes and 100 transfer pipettes. Each UPT contains 50μL of NAP Guard™ (≥ 742.5mM K2EDTA).
Qx Urine Preservative Transport
- Allows for an extended range of time and temperature conditions for specimen storage and transport
- Designed to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae
- For male and female urine specimens from symptomatic and asymptomatic individuals
- For use with the BD ProbeTec™ CT Qx and GC Qx Amplified DNA Assays on the BD Viper™ System in extracted mode
Specifications
Specifications
| Product Type | Urine Preservative Tube |
| Additive | NAP Guard™, 50μL |
| For Use With (Application) | For BD ProbeTec™ Chlamydia trachomatis and Neisseria gonorrhoeae Qx amplified DNA Assays, to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae in male and female Urine specimens |
| Includes | Each Qx UPT kit contains 100 Qx UPT tubes and 100 transfer pipettes, each UPT contains 50μL of NAP Guard™ |