missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Veal Infusion Broth

Product Code. 11748812
Click to view available options
:
Veal Infusion Broth; 500g
This item is not returnable. View return policy

Product Code. 11748812

Brand: BD 234420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivating fastidious microorganisms

  • Formulation: Lean Veal infusion from 500g 10g/L, No. 3 Proteose Peptone 10g/L, Sodium Chloride 5g/L

Order Info

Prices are for units of one each unless otherwise specified

Certifications

AOAC, BAM, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Veal Infusion Broth
Product Type Microbiology Media
Formulation Lean Veal infusion from 500g 10g/L, No. 3 Proteose Peptone 10g/L, Sodium Chloride 5g/L
For Use With (Application) Cultivation of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.