missing translation for 'onlineSavingsMsg'
Learn More

Beeswax, bleached, white, Thermo Scientific Chemicals

Product Code. 10411651 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10411651

Brand: Thermo Scientific Acros 222395000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Acidity 17 to 24mg KOH/g
CAS 8012-89-3
Color White to White-Yellow
Packaging Plastic bottle
MDL Number MFCD00132754
Quantity 500 g
Saponification Value 87 to 104mg KOH/g
Merck Index 15, 1019
Physical Form Pieces or Plates
Chemical Name or Material Beeswax

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.