missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ behrotest™ Workstation Equipment
Click to view available options
Sufficient For:
12 Samples
6 Samples
Unit Size:
Each
Description
- For use with samples of heavy metal digestion with aqua regia
Specifications
Specifications
| For Use With (Application) | twelve samples of heavy metal |
| Sufficient For | 12 Samples |
| Type | Behrotest workstation equipment |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction