Learn More
SP Bel-Art™ Bel Art Products Trypsin Warming Plate
Product Code. 15523034 Shop All SP Scienceware Products
Click to view available options
Dimensions (L x W x H):
25.4 x 24.9 x 5.7 cm
Unit Size:
Each
Product Code. 15523034
Brand: SP Bel-Art™ F370153000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Highly accurate Warming Plate, set at an optimal 37°C (±1°C), is designed to Trypsinize Cells during cell passaging
  • Avoids numerous entries into the CO2 incubator which alter its atmosphere, injuring ongoing experiments in the incubated cultures
  • Increases cell viability by keeping cells at an optimal temperature during examination of cell cultures outside of the incubator
  • Can be used to maintain the warmth of reagents inside the hood
  • Built in, 15 minute count down timer keeps track of the time that the trypsin has been on the cells
  • Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cm W (7 x 8 in.)
  • Fits easily in cell culture hoods or on the workbench
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant and safe for cleaning with Ethanol or IPA
Specifications
Temperature (Metric) 37°C
Width (Metric) 24.9 cm
Voltage 220 V
Dimensions (L x W x H) 25.4 x 24.9 x 5.7 cm