missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Benchtop Biohazard Waste Disposal Bag
Shop All SP Scienceware Products
Click to view available options
Color:
Red
Unit Size:
Pack of 100
Description
Bright red, autoclavable bags feature one long flap for easy opening and disposal of tips and tubes
- Size allows the bag to be inverted over user's hand to safely pick up waste
- Printed with the biohazard symbol and sterilization indicator patch
- Constructed from High Molecular Weight, High-Density (HMHD) polyethylene
- Steam autoclavable at 121°C (250°F)
- For space-saving waste disposal, use with Poxygrid™ Benchtop Biohazard Bag Holder (Mfr. No. H13193-1000)
Specifications
Specifications
| Design | Printed Biohazard Symbols |
| Capacity (English) | 0.4 gal. |
| Color | Red |
| Disposable | Single-use |
| Height (Metric) | 28 cm |
| Width (Metric) | 22 cm |
| Autoclavable | Autoclavable |
| For Use With (Application) | Easy Opening and Disposal of Tips and Tubes |
| Width (English) | 8.5 in. |
| Thickness | 0.07 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction