Learn More
Bel-Art™ Magnetic Stirring Bar Arrester
Shop All SP Scienceware Products
Click to view available options
Length (Metric):
51 mm
Unit Size:
Each
Description
- Radial magnet in thick polyethylene
Specifications
Specifications
| Diameter (English) | 2 in. |
| Dimensions (L x W x H) | 51 x 9.5 mm |
| For Use With (Application) | Prevent stirring bars from pouring out when emptying contents |
| For Use With (Equipment) | Stirring bars |
| Includes | Radial magnet |
| Thickness | 9.5mm |
| Diameter (Metric) | 5.1 cm |
| Length (Metric) | 51 mm |
| Material | Polyethylene |
| Quantity | 1 |
| Show More |