missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Reagent/Acid Pump Dispenser
Shop All SP Scienceware Products
Click to view available options
Autoclavable:
Not Autoclavable
Unit Size:
Each
Description
- Squeeze the small plastic bottle to begin flow; press the pressure release button to immediately stop dispensing
- Fits many 5 pint (80 fl. oz.) acid bottles, 1/2 and 1 gallon jugs, 2.5 and 4 liter bottles, and similar bottles with GPS No. 38-430 or 38-400 threaded necks (not included)
- Two adapters provided for short necks
Specifications
Specifications
| Angle | 45 degrees |
| Product Type | Reagent Dispenser |
| Autoclavable | Not Autoclavable |
| Color | White |
| Channels | 1 |
| Material | Polyethylene |
| Volume (Metric) | 4 L |
| For Use With (Application) | Dispense dangerous liquids without pouring |
| For Use With (Equipment) | 2.5, 4L, 5 pt., 0.5 gal., 1 gal. jugs and bottles |
| Includes | Siphon tube long enough to reach bottom of 2.4L Acid bottles |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction