Learn More
Bel Art™ Secador™ Desiccator Replacement Shelves
Shop All SP Scienceware Products
Click to view available options
Product Type:
Desiccator Shelf (Horizontal)
Desiccator Shelf (Vertical)
Unit Size:
Each
Description
- Fit auto and nonelectric models
- Durable, clear polystyrene construction
- Perforated to faciilitate air flow
Specifications
Specifications
| Diameter (Metric) Outer | 48 cm |
| Product Type | Desiccator Shelf (Horizontal) |
| Material | Polystyrene |
| For Use With (Application) | Convert 4.0 units from vertical to horizontal desiccator type |