Learn More
Bel-Art™ Spinbar™ Round Magnetic Stirring Bars
Product Code. 11366944 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
76.2 mm
101.6 mm
152.4 mm
Diameter (Metric):
12.7 mm
16 mm
19 mm
Length (English):
3 in.
4 in.
6 in.
Diameter (English):
0.5 in.
0.62 in.
0.75 in.
Unit Size:
Each
Product Code. 11366944
Brand: Bel-Art™ F371110003
Only null left
Add to basket
Add to basket
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
  • Small removable pivot ring in center adds to versatility
  • Pivot ring minimizes contact area of bar to vessel, reduces friction and lessens marring of plastic containers
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclave safe
Certifications
Manufactured in a registered ISO 9001:2008, as verified by SGS certification
Specifications
Autoclavable Autoclavable
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008
Color White
Diameter (English) 0.5 in.
Length (English) 3 in.
Coating PTFE, FDA-grade, USP Class VI
Diameter (Metric) 12.7 mm
Length (Metric) 76.2 mm
Material Alnico Magnet
Quantity 50
Shape Cylinder
Type Magnetic Stir Bar with Pivot Ring