Learn More
Bel-Art™ Spinbox™ Spinplus™ Magnetic Stirring Bar Assortment
Shop All SP Scienceware ProductsDescription
Includes
Octagon figured 16 assorted bars, standard and micro; standard bars with pivot ring Octagon shaped 12 assorted bars, standard bars with pivot rings Round shaped 22 assorted bars with removable pivot rings on bars longer than 1 in. Metric Polygon shaped 18 assorted bars Color-coded, octagon shaped 24 assorted bars with pivot ring Color-coded micro bars 12 assorted bars Egg shaped bars 13 assorted bars Spinplus in. + shaped bars 5 assorted bars
- Convenient storage box segregates sizes and styles for easy, no-rummage organization
- FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Available in octagon, egg and Spinplus™ shapes
- Autoclavable
Certifications
ISO 9001:2008
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | FDA, USP Class VI, ISO 9001:2008, SGS |
| Color | Red, Yellow, Blue |
| Includes | Two each of the following sizes: 0.5 x 0.31 in., 0.63 x 0.31 in., 1 x 0.38 in., 1.5 x 0.38 in., 2 x 0.38 in., and 2.5 x 0.38 in. |
| Coating | PTFE, FDA-grade, USP Class VI |
| Diameter (Metric) | 7.9 mm, 9.7 mm |
| Length (Metric) | 12.7 mm, 16 mm, 25.4 mm, 38.1 mm, 50.8 mm, 63.5 mm |
| Material | Alnico Magnet |
| Quantity | 6 |
| Shape | Octagon |
| Show More |