Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
Shop All SP Scienceware Products
Click to view available options
Unit Size:
Pack of 10
Pack of 100
Pack of 12
Capacity (Metric):
1.25 mL
2.5 mL
5 mL
8 mL
20 mL
Sterility:
Non-sterile
Sterile
Quantity:
10/Pk.
100/Pk.
12/Pk.
Description
- Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
- Flat-bottom shape permits use as a weighing boat
- For one-time use
Order Info
100 individually sterile wrapped spoons or 12 nonsterile spoons per pack
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Product Type | Spoon |