Learn More
Bel-Art™ Separator Discs
Product Code. 10013173 Shop All SP Scienceware Products
Click to view available options
Holds:
Tapes with 3 in. Core
Unit Size:
Pack of 3
Product Code. 10013173
Brand: Bel-Art™ F134690003
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For Write-On™ label tape dispensers.
  • Improve function of Write-On label tape dispensers
  • Use when loading with more than four rolls of labeling tape
Specifications
Product Type Separator Disc
Color White
Length (Metric) 76.2 mm
Width (Metric) 76.2 mm
Height (Metric) 76.2 mm
Holds Tapes with 3 in. Core
Material Plastic
Mounting Pre-drilled Holes
No. of Compartments 1
Product Style Side Loading
For Use With (Application) Improve the function of Write-On Label Tape Dispensers
For Use With (Equipment) For more than 4 rolls of labeling tape
Quantity 3/Pk.