Learn More
Emma Safety Footwear Bellagio Safety Shoes
A classically styled Brogue style Business shoe that always sets the right tone. Polished black full grain leather uppers and a virtually impenetrable nitrile rubber sole allows the wearer to stroll from the office to the factory and back again, with no fuss or shoe changing.
Brand: Emma Safety Footwear MM13709047
Additional Details : Weight : 1.87800kg
Description
- S3-Safety shoe with sole plate for wet work
- Nitrile rubber heat resistant outsole
- Full grain leather construction
- ESD certified
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
47 |
Laces | |
Bellagio | |
Leather | |
Steel |