Learn More
Emma Safety Footwear Bellagio XD Safety Shoes
Product Code. 15908231
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15908231
Brand: Emma Safety Footwear 13700041
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
A classically styled Brogue style Business shoe that always sets the right tone. Polished black full grain leather uppers and a virtually impenetrable nitrile rubber sole allows the wearer to stroll from the office to the factory and back again, with no fuss or shoe changing.
  • S3-Safety shoe with sole plate for wet work
  • Nitrile rubber heat resistant outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Design Bellagio XD
Material Leather
Size 41