Learn More
Kern Sohn Bench Scale GAB
Product Code. 17301663
Encompass_Preferred
Click to view available options
Capacity:
12 kg
15 kg, 30 kg
3 kg, 6 kg
30 kg
6 kg
6 kg, 15 kg
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17301663
Brand: Kern Sohn GAB 30K0.2N
Regulatory: This product is under eco-contribution tax.
Compact bench scale that is practical for small spaces.
  • Compact size, practical for small spaces
  • High mobility: thanks to rechargeable battery operation (optional), compact, lightweight construction, it is suitable for the use in several locations (production, warehouse, dispatch department etc.)
  • Weighing with tolerance range (check-weighing): a visual and audible signal helps with portioning, dispensing or grading
  • Totalising of weights
  • Protective working cover included with delivery
Specifications
Capacity 30 kg
Readability 0.2 g
Model Compact