Learn More
WTW Benchtop Meter inoLab Cond 7310P with built-in printer, EU & UK plugs
GREENER_CHOICE
Brand:  WTW 1CA300P
1707.24 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24121" to get your promotional price.
Benchtop Meter inoLab Cond 7310P with built-in printer, EU & UK plugs
Regulatory: This product is under eco-contribution tax.
Product Code. 15591746
  • ONLINE EXCLUSIVE PRICE
    2130.00€
    1879.00€ / Each
    Save 251.00€
    11% Off
Only null left
Estimated Shipment: 14-01-2026