Learn More
Benedict's Reagent, Pure, Qualitative, Fisher Chemical™
Product Code. 10325900 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Product Code. 10325900
Brand: Fisher Chemical J/2100/15
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Chemical Identifiers
MDL Number 146068
Specifications
Color Blue
Packaging Amber glass bottle
Quantity 1 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Benedict's reagent (qualitative)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium carbonate, Copper (II) sulfate pentahydrate (1:1:5)