missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benoxaprofen Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Benoxaprofen Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Benoxaprofen Ethyl Ester |
| CAS | 51234-41-4 |
| Analyte or Component Names | Benoxaprofen Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H16 Cl N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(4-Chlorophenyl)-alpha-methyl-5-benzoxazoleacetic acid ethyl ester |
| InChI Formula | InChI=1S/C18H16ClNO3/c1-3-22-18(21)11(2)13-6-9-16-15(10-13)20-17(23-16)12-4-7-14(19)8-5-12/h4-11H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(C)c1ccc2oc(nc2c1)c3ccc(Cl)cc3 |
| IUPAC Name | ethyl 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction