Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Specifications
| Type | Bent Receiver Adapter with Glass Olive |
| Material | Duran Glass |
| Certifications/Compliance | DIN 12594 |
| Standard Taper Joint | NS 24/29 |