Learn More
Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals
Catalog Number p-62723
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
1125-88-8
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC
1125-88-8
MFCD00008491
HEVMDQBCAHEHDY-UHFFFAOYSA-N
benzaldehyde dimethyl acetal, dimethoxymethyl benzene, alpha,alpha-dimethoxytoluene, dimethoxyphenylmethane, benzene, dimethoxymethyl, benzaldehydedimethylacetal, dimethoxymethyl-benzene, benzaldehyde, dimethyl acetal, benzaldehyde dimethylacetal, unii-w6t83k35ca
62375
dimethoxymethylbenzene
COC(C1=CC=CC=C1)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H12O2
Linear Formula C6H5CH(OCH3)2
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Quantity 5 mL
Molecular Weight (g/mol) 152.193
Formula Weight 152.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Benzaldehyde dimethyl acetal, 99+%